siRNA Details
| Gene Symbol: Tie1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053545.1 | NP_445997.1 | 11, 12 | 1676 | ||
| XM_006238720.2 | XP_006238782.1 | 11, 12 | 1744 | |||
| GenBank mRNA | BC169020.1 | - | 11, 12 | 1676 | 9099086 | |

Tie1
Genomic Map