siRNA Details
Gene Symbol: Pdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053551.1 | NP_446003.1 | 11 | 1279 | ||
GenBank mRNA | AF034577.1 | - | 11 | 1279 | ||
FQ216954.1 | - | 1322 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053551.1 | NP_446003.1 | 11 | 1279 | ||
GenBank mRNA | AF034577.1 | - | 11 | 1279 | ||
FQ216954.1 | - | 1322 |