siRNA Details
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048180.1 | NP_001041645.1 | 6 | 1689 | ||
NM_134456.3 | NP_604451.2 | 7 | 1989 | |||
XM_006230351.2 | XP_006230413.1 | 6 | 3550 | |||
XM_006230352.2 | XP_006230414.1 | 6 | 3550 | |||
XM_006230353.2 | XP_006230415.1 | 7 | 1964 | |||
XM_006230354.2 | XP_006230416.1 | 7 | 2870 | |||
XM_006230355.2 | XP_006230417.1 | 6 | 3549 | |||
GenBank mRNA | AF047577.1 | - | 6 | 1689 | ||
U57391.1 | - | 7 | 2032 |
