siRNA Details
| Gene Symbol: Trpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053559.1 | NP_446011.1 | 11 | 2520 | ||
| GenBank mRNA | FQ212829.1 | - | 5 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpc6| Gene Symbol: Trpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053559.1 | NP_446011.1 | 11 | 2520 | ||
| GenBank mRNA | FQ212829.1 | - | 5 | 687 | ||