siRNA Details
| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053567.1 | NP_446019.1 | 6 | 729 | ||
| GenBank mRNA | AF079233.3 | - | 6 | 729 | ||
| BC091134.1 | - | 6 | 741 | 7369085 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ftcd| Gene Symbol: Ftcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053567.1 | NP_446019.1 | 6 | 729 | ||
| GenBank mRNA | AF079233.3 | - | 6 | 729 | ||
| BC091134.1 | - | 6 | 741 | 7369085 | ||