siRNA Details
| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053575.2 | NP_446027.2 | 25 | 4185 | ||
| GenBank mRNA | AF061947.1 | - | 25 | 4191 | ||
| Gene Symbol: LOC103689948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256337.2 | XP_006256399.1 | 1326 | |||

Cabin1
Genomic Map