siRNA Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053582.2 | NP_446034.1 | 6 | 723 | ||
| XM_006238977.2 | XP_006239039.1 | 7 | 815 | |||
| XM_006238978.2 | XP_006239040.1 | 7 | 812 | |||
| GenBank mRNA | AB050717.1 | - | 6 | 724 | ||
| BC097299.1 | - | 7 | 782 | 7445067 | ||

Tinagl1
Genomic Map