siRNA Details
| Gene Symbol: Dpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053591.2 | NP_446043.1 | 8 | 934 | ||
| XM_006255783.2 | XP_006255845.1 | 8 | 929 | |||
| GenBank mRNA | BC072476.1 | - | 8 | 934 | 7100683 | |
| L07315.1 | - | 8 | 912 | |||
| L07316.1 | - | 8 | 912 | |||
| M94056.1 | - | 8 | 912 | |||

Dpep1
Genomic Map