siRNA Details
| Gene Symbol: Ptprr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113390.1 | NP_001106861.1 | 10 | 1364 | ||
| NM_053594.2 | NP_446046.2 | 2073 | ||||
| GenBank mRNA | BC089186.1 | - | 6 | 855 | 7315399 | |
| D38292.1 | - | 2076 | ||||
| D64050.1 | - | 10 | 1392 | |||
| U14914.1 | - | 1420 | ||||

Ptprr
Genomic Map