siRNA Details
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080778.2 | NP_542956.1 | 2 | 716 | ||
XM_006229366.2 | XP_006229428.1 | 2 | 5603 | |||
GenBank mRNA | AF003944.1 | - | 2 | 717 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080778.2 | NP_542956.1 | 2 | 716 | ||
XM_006229366.2 | XP_006229428.1 | 2 | 5603 | |||
GenBank mRNA | AF003944.1 | - | 2 | 717 |