siRNA Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053643.1 | NP_446095.1 | 10 | 940 | ||
| GenBank mRNA | AB052898.1 | - | 10 | 940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cds2| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053643.1 | NP_446095.1 | 10 | 940 | ||
| GenBank mRNA | AB052898.1 | - | 10 | 940 | ||