siRNA Details
| Gene Symbol: Kremen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053649.1 | NP_446101.1 | 6 | 691 | ||
| GenBank mRNA | AB065090.1 | - | 6 | 691 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kremen1| Gene Symbol: Kremen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053649.1 | NP_446101.1 | 6 | 691 | ||
| GenBank mRNA | AB065090.1 | - | 6 | 691 | ||