siRNA Details
| Gene Symbol: Flt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053652.1 | NP_446104.1 | 8 | 1069 | ||
| GenBank mRNA | AF402785.1 | - | 8 | 1069 | ||
| AF402786.1 | - | 8 | 1069 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flt4| Gene Symbol: Flt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053652.1 | NP_446104.1 | 8 | 1069 | ||
| GenBank mRNA | AF402785.1 | - | 8 | 1069 | ||
| AF402786.1 | - | 8 | 1069 | |||