siRNA Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053707.2 | NP_446159.1 | 2 | 390 | ||
| GenBank mRNA | AF389348.1 | - | 2 | 119 | ||
| AF448810.1 | - | 2 | 119 | |||
| BC070943.1 | - | 2 | 390 | 7103046 | ||
| FQ231256.1 | - | 3 | 153 | |||
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ007291.1 | - | 1004 | |||

Hdgf
Genomic Map