siRNA Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053707.2 | NP_446159.1 | 5 | 970 | ||
| GenBank mRNA | AF389348.1 | - | 5 | 699 | ||
| BC070943.1 | - | 5 | 970 | 7103046 | ||
| FQ231256.1 | - | 6 | 733 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdgf| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053707.2 | NP_446159.1 | 5 | 970 | ||
| GenBank mRNA | AF389348.1 | - | 5 | 699 | ||
| BC070943.1 | - | 5 | 970 | 7103046 | ||
| FQ231256.1 | - | 6 | 733 | |||