siRNA Details
| Gene Symbol: Bcl2l10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053733.1 | NP_446185.1 | 2 | 519 | ||
| GenBank mRNA | AY029163.1 | - | 2 | 519 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl2l10| Gene Symbol: Bcl2l10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053733.1 | NP_446185.1 | 2 | 519 | ||
| GenBank mRNA | AY029163.1 | - | 2 | 519 | ||