siRNA Details
| Gene Symbol: Trafd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053760.2 | NP_446212.2 | 6 | 932 | ||
| XM_006249376.2 | XP_006249438.1 | 6 | 988 | |||
| XM_008769209.1 | XP_008767431.1 | 6 | 1035 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trafd1| Gene Symbol: Trafd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053760.2 | NP_446212.2 | 6 | 932 | ||
| XM_006249376.2 | XP_006249438.1 | 6 | 988 | |||
| XM_008769209.1 | XP_008767431.1 | 6 | 1035 | |||