siRNA Details
| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007602.1 | NP_001007603.1 | 6 | 747 | ||
| GenBank mRNA | AB008807.1 | - | 6 | 729 | ||
| BC079363.1 | - | 6 | 747 | 7109248 | ||
| FJ179405.1 | - | 6 | 665 | |||
| FQ218105.1 | - | 6 | 722 | |||
| FQ220742.1 | - | 6 | 689 | |||

Gsto1
Genomic Map