siRNA Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053776.2 | NP_446228.2 | 4 | 548 | ||
| XM_008762628.1 | XP_008760850.1 | 5 | 857 | |||
| XM_008762629.1 | XP_008760851.1 | 2 | 110 | |||
| GenBank mRNA | AF118853.1 | - | 4 | 521 | ||
| AY322161.1 | - | 4 | 521 | |||
| AY322162.1 | - | 4 | 521 | |||
| AY322163.1 | - | 4 | 432 | |||
| BC088838.1 | - | 4 | 548 | 7132881 | ||

Dnajc2
Genomic Map