siRNA Details
| Gene Symbol: Stx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053788.2 | NP_446240.2 | 5, 6 | 398 | ||
| GenBank mRNA | AF217191.1 | - | 5, 6 | 367 | ||
| D10392.1 | - | 5, 6 | 398 | |||
| D12519.1 | - | 5, 6 | 358 | |||
| M95734.1 | - | 5, 6 | 359 | |||

Stx1a
Genomic Map