siRNA Details
| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053796.1 | NP_446248.1 | 5 | 556 | ||
| GenBank mRNA | AF241261.1 | - | 1 | 103 | ||
| AF276998.1 | - | 5 | 556 | |||
| BC065309.1 | - | 5 | 538 | 5599898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F11r| Gene Symbol: F11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053796.1 | NP_446248.1 | 5 | 556 | ||
| GenBank mRNA | AF241261.1 | - | 1 | 103 | ||
| AF276998.1 | - | 5 | 556 | |||
| BC065309.1 | - | 5 | 538 | 5599898 | ||