siRNA Details
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053835.1 | NP_446287.1 | 5 | 524 | ||
GenBank mRNA | FQ213983.1 | - | 5 | 614 | ||
M15883.1 | - | 5 | 524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053835.1 | NP_446287.1 | 5 | 524 | ||
GenBank mRNA | FQ213983.1 | - | 5 | 614 | ||
M15883.1 | - | 5 | 524 |