siRNA Details
| Gene Symbol: Il2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053836.1 | NP_446288.1 | 3 | 328 | ||
| GenBank mRNA | M22899.1 | - | 3 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il2| Gene Symbol: Il2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053836.1 | NP_446288.1 | 3 | 328 | ||
| GenBank mRNA | M22899.1 | - | 3 | 328 | ||