siRNA Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169113.1 | NP_001162584.1 | 4 | 585 | ||
| GenBank mRNA | BC082020.1 | - | 4 | 553 | ||
| BC098947.1 | - | 4 | 550 | |||
| M31051.1 | - | 4 | 432 | |||
| Z11994.1 | - | 545 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrpap1| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169113.1 | NP_001162584.1 | 4 | 585 | ||
| GenBank mRNA | BC082020.1 | - | 4 | 553 | ||
| BC098947.1 | - | 4 | 550 | |||
| M31051.1 | - | 4 | 432 | |||
| Z11994.1 | - | 545 | ||||