siRNA Details
| Gene Symbol: Cap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053874.1 | NP_446326.1 | 12 | 1361 | ||
| XM_006253753.2 | XP_006253815.1 | 13 | 1470 | |||
| XM_006253754.2 | XP_006253816.1 | 13 | 1583 | |||
| GenBank mRNA | U31935.1 | - | 12 | 1361 | ||

Cap2
Genomic Map