siRNA Details
Gene Symbol: Cyth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053910.1 | NP_446362.1 | 11 | 944 | ||
XM_006247784.2 | XP_006247846.1 | 10 | 985 | |||
XM_006247785.2 | XP_006247847.1 | 11 | 2079 | |||
XM_008768306.1 | XP_008766528.1 | 10 | 3384 | |||
XM_008768307.1 | XP_008766529.1 | 10 | 941 | |||
GenBank mRNA | BC166417.1 | - | 10 | 941 | 9028626 | |
U83895.1 | - | 11 | 944 |
