siRNA Details
| Gene Symbol: Acacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053922.1 | NP_446374.1 | 34 | 4668 | ||
| XM_008769309.1 | XP_008767531.1 | 35 | 5169 | |||
| XM_008769310.1 | XP_008767532.1 | 35 | 5773 | |||
| XM_008769311.1 | XP_008767533.1 | 36 | 5278 | |||
| XM_008769312.1 | XP_008767534.1 | 35 | 5264 | |||
| XM_008769313.1 | XP_008767535.1 | 35 | 5148 | |||
| XM_008769314.1 | XP_008767536.1 | 34 | 4524 | |||
| GenBank mRNA | AB004329.1 | - | 34 | 4668 | ||
| DQ493871.1 | - | 34 | 4688 | |||

Acacb
Genomic Map