siRNA Details
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053925.1 | NP_446377.1 | 449 | |||
GenBank mRNA | AB022014.1 | - | 449 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053925.1 | NP_446377.1 | 449 | |||
GenBank mRNA | AB022014.1 | - | 449 |