siRNA Details
| Gene Symbol: Kcnh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053937.1 | NP_446389.1 | 8 | 1971 | ||
| XM_006247554.2 | XP_006247616.1 | 8 | 1957 | |||
| XM_006247555.2 | XP_006247617.1 | 8 | 1957 | |||
| XM_006247556.2 | XP_006247618.1 | 8 | 1957 | |||
| XM_008768308.1 | XP_008766530.1 | 8 | 1957 | |||
| XM_008768309.1 | XP_008766531.1 | 8 | 1957 | |||
| GenBank mRNA | AF016192.1 | - | 8 | 1971 | ||

Kcnh6
Genomic Map