siRNA Details
| Gene Symbol: Kcns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053954.1 | NP_446406.1 | 3 | 1039 | ||
| XM_006235509.2 | XP_006235571.1 | 2 | 1455 | |||
| GenBank mRNA | Y17606.1 | - | 3 | 1039 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcns1| Gene Symbol: Kcns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053954.1 | NP_446406.1 | 3 | 1039 | ||
| XM_006235509.2 | XP_006235571.1 | 2 | 1455 | |||
| GenBank mRNA | Y17606.1 | - | 3 | 1039 | ||