siRNA Details
Gene Symbol: Rhbdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191822.1 | NP_001178751.1 | 6 | 757 | ||
XM_006245894.2 | XP_006245956.1 | 5 | 2337 | |||
XM_008767537.1 | XP_008765759.1 | 6 | 2511 | |||
GenBank mRNA | Y17258.1 | - | 3 | 141 |
