siRNA Details
| Gene Symbol: Mmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133514.1 | NP_598198.1 | 7 | 1008 | ||
| GenBank mRNA | M65253.1 | - | 7 | 1008 | ||
| X05083.1 | - | 7 | 1005 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp10| Gene Symbol: Mmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133514.1 | NP_598198.1 | 7 | 1008 | ||
| GenBank mRNA | M65253.1 | - | 7 | 1008 | ||
| X05083.1 | - | 7 | 1005 | |||