siRNA Details
| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057142.1 | NP_476483.1 | 18 | 1927 | ||
| GenBank mRNA | EU348630.1 | - | 4 | 494 | ||
| EU348638.1 | - | 3 | 263 | |||
| U66707.1 | - | 18 | 1927 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc7| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057142.1 | NP_476483.1 | 18 | 1927 | ||
| GenBank mRNA | EU348630.1 | - | 4 | 494 | ||
| EU348638.1 | - | 3 | 263 | |||
| U66707.1 | - | 18 | 1927 | |||