siRNA Details
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008767.1 | NP_001008767.1 | 5 | 874 | ||
GenBank mRNA | BC088411.1 | - | 5 | 874 | 7192327 | |
U30789.1 | - | 5 | 1659 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008767.1 | NP_001008767.1 | 5 | 874 | ||
GenBank mRNA | BC088411.1 | - | 5 | 874 | 7192327 | |
U30789.1 | - | 5 | 1659 |