siRNA Details
| Gene Symbol: Keap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057152.2 | NP_476493.2 | 2 | 806 | ||
| XM_006242591.2 | XP_006242653.1 | 2 | 840 | |||
| XM_006242592.2 | XP_006242654.1 | 1 | 1341 | |||
| GenBank mRNA | AF304364.1 | - | 1 | 560 | ||
| FQ212383.1 | - | 2 | 806 | |||

Keap1
Genomic Map