siRNA Details
| Gene Symbol: Gmpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057188.1 | NP_476536.1 | 4 | 398 | ||
| GenBank mRNA | AF090867.1 | - | 4 | 398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmpr| Gene Symbol: Gmpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057188.1 | NP_476536.1 | 4 | 398 | ||
| GenBank mRNA | AF090867.1 | - | 4 | 398 | ||