siRNA Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080584.4 | NP_542151.1 | 9 | 1338 | ||
| XM_008759855.1 | XP_008758077.1 | 10 | 1161 | |||
| XM_008759856.1 | XP_008758078.1 | 10 | 1279 | |||
| GenBank mRNA | BC107906.1 | - | 9 | 1198 | ||
| BC128715.1 | - | 9 | 1267 | 7456955 | ||
| M73808.1 | - | 9 | 1338 | |||

Phkg2
Genomic Map