siRNA Details
| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080896.1 | NP_543172.1 | 9 | 1013 | ||
| GenBank mRNA | AF367468.1 | - | 9 | 1013 | ||
| FQ212957.1 | - | 6 | 592 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021510, CO428102, AK165283, AK161491, BC056224, and others.

Hnrnph1
Genomic Map