siRNA Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130429.1 | NP_569113.1 | 9 | 1027 | ||
| XM_006233302.2 | XP_006233364.1 | 9 | 2346 | |||
| XM_006233303.2 | XP_006233365.1 | 9 | 2345 | |||
| XM_006233305.2 | XP_006233367.1 | 8 | 2262 | |||
| XM_006233306.2 | XP_006233368.1 | 8 | 2261 | |||
| XM_006233309.2 | XP_006233371.1 | 9 | 1779 | |||
| XM_008761489.1 | XP_008759711.1 | 9 | 2348 | |||
| XM_008761490.1 | XP_008759712.1 | 8 | 2264 | |||
| XM_008761491.1 | XP_008759713.1 | 9 | 1042 | |||
| XM_008761492.1 | XP_008759714.1 | 9 | 1779 | |||
| GenBank mRNA | AF198533.1 | - | 9 | 1027 | ||

Lef1
Genomic Map