siRNA Details
| Gene Symbol: Rasgrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130824.2 | NP_570837.2 | 14 | 1773 | ||
| GenBank mRNA | AF465263.1 | - | 14 | 1773 | ||
| AF465264.1 | - | 14 | 1776 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgrp4| Gene Symbol: Rasgrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130824.2 | NP_570837.2 | 14 | 1773 | ||
| GenBank mRNA | AF465263.1 | - | 14 | 1773 | ||
| AF465264.1 | - | 14 | 1776 | |||