siRNA Details
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133310.2 | NP_579844.2 | 5 | 820 | ||
GenBank mRNA | AF075595.1 | - | 5 | 608 | ||
BC083923.1 | - | 6 | 1025 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133310.2 | NP_579844.2 | 5 | 820 | ||
GenBank mRNA | AF075595.1 | - | 5 | 608 | ||
BC083923.1 | - | 6 | 1025 |