siRNA Details
| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133310.2 | NP_579844.2 | 5 | 859 | ||
| GenBank mRNA | AF075595.1 | - | 5 | 647 | ||
| BC083923.1 | - | 6 | 1064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icmt| Gene Symbol: Icmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133310.2 | NP_579844.2 | 5 | 859 | ||
| GenBank mRNA | AF075595.1 | - | 5 | 647 | ||
| BC083923.1 | - | 6 | 1064 | |||