siRNA Details
| Gene Symbol: Il24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133311.1 | NP_579845.1 | 6 | 712 | ||
| XM_008769421.1 | XP_008767643.1 | 5 | 1694 | |||
| GenBank mRNA | AF004774.2 | - | 6 | 661 | ||
| AF269251.1 | - | 6 | 712 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il24| Gene Symbol: Il24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133311.1 | NP_579845.1 | 6 | 712 | ||
| XM_008769421.1 | XP_008767643.1 | 5 | 1694 | |||
| GenBank mRNA | AF004774.2 | - | 6 | 661 | ||
| AF269251.1 | - | 6 | 712 | |||