siRNA Details
| Gene Symbol: Slc25a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133418.1 | NP_596909.1 | 2 | 202 | ||
| GenBank mRNA | AJ223355.1 | - | 2 | 202 | ||
| BC081734.1 | - | 2 | 220 | 7108552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a10| Gene Symbol: Slc25a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133418.1 | NP_596909.1 | 2 | 202 | ||
| GenBank mRNA | AJ223355.1 | - | 2 | 202 | ||
| BC081734.1 | - | 2 | 220 | 7108552 | ||