siRNA Details
| Gene Symbol: Chrna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133420.1 | NP_596911.1 | 7 | 1765 | ||
| XM_008770735.1 | XP_008768957.1 | 7 | 1607 | |||
| GenBank mRNA | AY574253.1 | - | 6 | 1461 | ||
| L10077.1 | - | 7 | 1765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna2| Gene Symbol: Chrna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133420.1 | NP_596911.1 | 7 | 1765 | ||
| XM_008770735.1 | XP_008768957.1 | 7 | 1607 | |||
| GenBank mRNA | AY574253.1 | - | 6 | 1461 | ||
| L10077.1 | - | 7 | 1765 | |||