siRNA Details
| Gene Symbol: Csap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133622.2 | NP_598306.2 | 467 | |||
| GenBank mRNA | BI284022.1 | - | 1 | 137 | ||
| U00964.1 | - | 467 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csap1| Gene Symbol: Csap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133622.2 | NP_598306.2 | 467 | |||
| GenBank mRNA | BI284022.1 | - | 1 | 137 | ||
| U00964.1 | - | 467 | ||||