siRNA Details
Gene Symbol: Csap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133622.2 | NP_598306.2 | 362 | |||
GenBank mRNA | BI284022.1 | - | 1 | 242 | ||
U00964.1 | - | 362 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133622.2 | NP_598306.2 | 362 | |||
GenBank mRNA | BI284022.1 | - | 1 | 242 | ||
U00964.1 | - | 362 |