siRNA Details
Gene Symbol: Gp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134418.3 | NP_599245.2 | 3 | 464 | ||
XM_008759672.1 | XP_008757894.1 | 3 | 460 | |||
GenBank mRNA | M58716.1 | - | 3 | 466 | ||
X53935.1 | - | 2 | 365 |
