siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134461.1 | NP_604456.1 | 2 | 128 | ||
| GenBank mRNA | AF484958.1 | - | 2 | 128 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn5| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134461.1 | NP_604456.1 | 2 | 128 | ||
| GenBank mRNA | AF484958.1 | - | 2 | 128 | ||