siRNA Details
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138518.2 | NP_612527.2 | 4 | 656 | ||
XM_006255708.2 | XP_006255770.1 | 5 | 779 | |||
GenBank mRNA | AF109674.1 | - | 4 | 671 | ||
CB732240.1 | - | 2 | 248 |
